Learn More
BINDER™ Telescopic Rack
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB Series
KB/KB PRO 260, KB/KB PRO 470, KBF/KBF PRO 260
KBF PRO Series
KBF Series
Unit Size:
Each
Description
- Shelf rack made of stainless steel
- Includes set of telescopic rails
Specifications
Specifications
| For Use With (Equipment) | KB/KB PRO 260, KB/KB PRO 470, KBF/KBF PRO 260 |
| Material | Stainless Steel |
| Type | Rack |