Learn More
Buerkle™ Telescopic Rod
Telescopic Rod is a versatile sampling system with exchangeable tools for a wide range of applications.
£51.65 - £190.00
Product Code | Brand | Length (Metric) | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Includes | Price | Quantity & Availability | |||||
10396242
|
Buerkle™
5355-0120 |
65 to 120 cm (Adjustable) | 2 Rods |
£54.00
Each |
In Stock
Log in to see stock. |
|||||
10530053
|
Buerkle™
5355-0250 |
125 to 250 cm | 2 Rods |
£70.70
Each |
In Stock
Log in to see stock. |
|||||
15376297
|
Buerkle™
5355-0180 |
70 to 180 cm | 4 Rods |
£51.65
Each |
In Stock
Log in to see stock. |
|||||
10376292
|
Buerkle™
5355-0280 |
95 to 280 cm (Adjustable) | 4 Rods |
£73.60
Each |
In Stock
Log in to see stock. |
|||||
10040822
|
Buerkle™
5355-0300 |
115 to 300 cm (Adjustable) | 3 Rods |
£106.00
Each |
In Stock
Log in to see stock. |
|||||
10182412
|
Buerkle™
5355-0450 |
165 to 450 cm (Adjustable) | 3 Rods |
£148.00
Each |
In Stock
Log in to see stock. |
|||||
10244992
|
Buerkle™
5355-0600 |
175 to 600 cm (Adjustable) | 4 Rods |
£190.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- Different tools can be attached to the telescopic rod
- The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint
- Ideal for sampling from baths, tanks, shafts, open water courses etc
- Sampling depths down to 6 m can be reached with the adjustable telescopic aluminum rod
- The telescopic rods cannot be rotated radially, which means even better handling
- The individual rods have been provided with an end stop, this means that one rod cannot accidentally slip out of another rod
- Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm. Fits into every car boot!
- Springs made of stainless steel
Specifications
Telescopic Rod | |
Aluminum |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Buerkle™ Telescopic Rod