missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Filling Tube for Dispensette™ S / S Organic
Manufactured from FEP, these Telescoping Filling Tubes adjust to various bottle heights.
£19.76 - £26.65
Product Code | Brand | Diameter (Metric) Outer Neck | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer Neck | Length (Metric) | Price | Quantity & Availability | ||||
15408023
|
BRAND™
708212 |
6 mm | From 125 to 240 mm |
£19.76
Each |
In Stock
Log in to see stock. |
||||
15438023
|
BRAND™
708218 |
7.6 mm | From 170 to 330 mm |
£26.65
Each |
In Stock
Log in to see stock. |
||||
Description
- FEP
Specifications
FEP | |
1/Pk. |