missing translation for 'onlineSavingsMsg'
Learn More
Tellurium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals Product Code.: 10098219

Tellurium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals

Product Code. 10098219
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10098219

Brand: Thermo Scientific Acros 437011000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compoundsShow MoreShow Less
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Boiling Point 990.0°C
Melting Point 450.0°C
Color Black to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.999%
Merck Index 15,9263
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in h2so4, hno3 and koh, insoluble in benzene and carbon disulfide
Formula Weight 127.6
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Tellurium
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465392 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number437011000,437010250
Lot NumberA0443226 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number437011000,437010250
Lot NumberA0437915 Certificate TypeCertificate of Analysis Date12/01/2023 Catalog Number437011000,437010250
Lot NumberA0395736 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number437011000,437010250
Lot NumberA0398289 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number437011000,437010250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tellurium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.