Learn More
Tellurium(IV) iodide, 99% (metals basis), Thermo Scientific Chemicals
£49.10 - £243.00
Chemical Identifiers
CAS | 7790-48-9 |
---|---|
Molecular Formula | I4Te |
Molecular Weight (g/mol) | 635.218 |
MDL Number | MFCD00049578 |
InChI Key | XCOKHDCPVWVFKS-UHFFFAOYSA-N |
Synonym | tellurium tetraiodide, tellurium iv iodide, tellurium iodide tei4 , t-4, unii-zb48i033v5, tetraiodo-, e4-tellane, tei4, tellurium iv iodide-te |
PubChem CID | 82255 |
SMILES | [Te](I)(I)(I)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11345039
|
Thermo Scientific Alfa Aesar
087795.06 |
5 g |
£49.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355039
|
Thermo Scientific Alfa Aesar
087795.14 |
25 g |
£243.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tellurium(IV) iodide is used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-48-9 | |
635.218 | |
XCOKHDCPVWVFKS-UHFFFAOYSA-N | |
82255 |
I4Te | |
MFCD00049578 | |
tellurium tetraiodide, tellurium iv iodide, tellurium iodide tei4 , t-4, unii-zb48i033v5, tetraiodo-, e4-tellane, tei4, tellurium iv iodide-te | |
[Te](I)(I)(I)I |
Specifications
(decomposition) | |
7790-48-9 | |
I4Te | |
UN2923 | |
tellurium tetraiodide, tellurium iv iodide, tellurium iodide tei4 , t-4, unii-zb48i033v5, tetraiodo-, e4-tellane, tei4, tellurium iv iodide-te | |
XCOKHDCPVWVFKS-UHFFFAOYSA-N | |
635.218 | |
635.22 | |
Acrid | |
5.403 g/mL at 15°C | |
Tellurium (IV) iodide |
280°C | |
Crystalline | |
MFCD00049578 | |
14,9127 | |
Decomposes slowly in cold water to TeO2 and HI | |
[Te](I)(I)(I)I | |
82255 | |
99% | |
Moisture sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 232-210-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only