missing translation for 'onlineSavingsMsg'
Learn More
Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals Product Code.: 10793411

Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals

Product Code. 10793411
250 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10793411

Brand: Thermo Scientific Acros 222592500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-07-3
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4Show MoreShow Less
PubChem CID 24007
IUPAC Name (oxo-λ⁴-selanylidene)oxidane
SMILES O=[Se]=O

Specifications

Boiling Point 1245°C
Melting Point 733°C
Color White to Yellow
Physical Form Powder
Quantity 250 g
Assay Percent Range 99+%
Linear Formula TeO2
Merck Index 15, 9265
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. acids and alkalies,slightly soluble in diluted acids
Formula Weight 159.6
Percent Purity ≥99%
Packaging Glass bottle
Density 5.67g/mL
Chemical Name or Material Tellurium(IV) oxide, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457596 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number222590500,222592500
Lot NumberA0454926 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number222590500,222592500
Lot NumberA0449560 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog Number222590500,222592500
Lot NumberA0394846 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number222590500,222592500
Lot NumberA0416717 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number222590500,222592500
Product Identifier
  • Tellurium(IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.