missing translation for 'onlineSavingsMsg'
Learn More

Tellurium lump, 99.999+% (metals basis), Thermo Scientific Chemicals

Product Code. 11345818
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11345818

Brand: Thermo Scientific Alfa Aesar 010362.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compoundsShow MoreShow Less
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Quantity 25 g
Odor Odorless
Form Lump
Percent Purity ≥99.999%
Assay (metals basis)
Chemical Name or Material Tellurium lump

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberR19L004 Certificate TypeCertificate of Analysis Date20/05/2025 Product Code010362.14, 010362.36, 010362.22
Lot NumberY15K005 Certificate TypeCertificate of Analysis Date19/11/2024 Product Code010362.14, 010362.36, 010362.22
Lot NumberT17K002 Certificate TypeCertificate of Analysis Date23/07/2024 Product Code010362.14, 010362.36, 010362.22
Lot NumberM16K028 Certificate TypeCertificate of Analysis Date23/01/2024 Product Code010362.14, 010362.36, 010362.22
Lot NumberM20I016 Certificate TypeCertificate of Analysis Date12/04/2022 Product Code010362.14, 010362.36, 010362.22
compliance-icons compliance-icons
Product Identifier
  • Tellurium lump
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H362-May cause harm to breast-fed children.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tellurium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tellurium lump, 99.999+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.