missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tellurium lump, typically 12mm (0.47 in.), 99.9999% (metals basis), Thermo Scientific Chemicals
£45.41 - £791.00
Chemical Identifiers
CAS | 13494-80-9 |
---|---|
Molecular Formula | Te |
Molecular Weight (g/mol) | 127.60 |
MDL Number | MFCD00134062 |
InChI Key | PORWMNRCUJJQNO-UHFFFAOYSA-N |
Synonym | tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds |
PubChem CID | 6327182 |
ChEBI | CHEBI:30452 |
IUPAC Name | tellurium |
SMILES | [Te] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306857
|
Thermo Scientific Alfa Aesar
010758.09 |
10 g | |||||||
11316857
|
Thermo Scientific Alfa Aesar
010758.18 |
50 g |
£184.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396847
|
Thermo Scientific Alfa Aesar
010758.30 |
250 g |
£791.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13494-80-9 | |
127.60 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
6327182 | |
tellurium |
Te | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
CHEBI:30452 | |
[Te] |
Specifications
13494-80-9 | |
MFCD00134062 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
tellurium | |
6327182 | |
99.9999% | |
Lump | |
Tellurium lump, Typically 12mm (0.47 in.) |
Te | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
[Te] | |
127.60 | |
CHEBI:30452 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'einecsNumber' : 236-813-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only