missing translation for 'onlineSavingsMsg'
Learn More

Tellurium powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11356088
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
13494-80-9
Te
127.60
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
6327182
CHEBI:30452
tellurium
[Te]
This item is not returnable. View return policy
13494-80-9
Te
127.60
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
6327182
CHEBI:30452
tellurium
[Te]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.999% (Metals basis)
Odor Odorless
Appearance Gray powder
Chemical Name or Material Tellurium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'