missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEMED (Electrophoresis), Fisher BioReagents
For polymerization of acrylamide/bis-acrylamide
£61.40 - £111.00
Chemical Identifiers
CAS | 110-18-9 |
---|---|
Molecular Formula | C6H16N2 |
Molecular Weight (g/mol) | 116.208 |
InChI Key | KWYHDKDOAIKMQN-UHFFFAOYSA-N |
Synonym | temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl |
PubChem CID | 8037 |
ChEBI | CHEBI:32850 |
IUPAC Name | N,N,N',N'-tetramethylethane-1,2-diamine |
SMILES | CN(C)CCN(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10689543
|
Fisher BioReagents
BP150-20 |
26 mL | Amber Glass |
£61.40
20g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10142863
|
Fisher BioReagents
BP150-100 |
130 mL | Amber Glass |
£111.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TEMED is a commonly used catalyst for the polymerization of acrylamide/bis-acrylamide.Chemical Identifiers
110-18-9 | |
116.208 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CHEBI:32850 | |
CN(C)CCN(C)C |
C6H16N2 | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
8037 | |
N,N,N',N'-tetramethylethane-1,2-diamine |
Specifications
110-18-9 | |
Colorless | |
8.0 to 8.5 | |
≥97 % (by titration) | |
(CH3)2NCH2CH2N(CH3)2 | |
15, 9277 | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylethane-1,2-diamine | |
8037 | |
116.21 | |
DNase-, RNase- and Protease-Free | |
TEMED |
-55°C | |
0.770g/cm³ | |
120°C | |
C6H16N2 | |
2372 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CN(C)CCN(C)C | |
116.208 | |
CHEBI:32850 | |
≥97% | |
Liquid |
Safety and Handling
missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid
Recommended Storage : RT
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- N,N,N',N'-Tetramethylethylenediamine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P240-Ground/bond container and receiving equipment.