missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEMED, PlusOne™, Cytiva
Click to view available options
Quantity:
25 mL
Specifications
CAS | 110-18-9 |
Assay Percent Range | ≥99% |
Molecular Formula | C6H16N2 |
Quantity | 25 mL |
Synonym | temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl |
InChI Key | KWYHDKDOAIKMQN-UHFFFAOYSA-N |
SMILES | CN(C)CCN(C)C |
IUPAC Name | N,N,N',N'-tetramethylethane-1,2-diamine |
Molecular Weight (g/mol) | 116.208 |
PubChem CID | 8037 |
Show More |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305-IF IN EYES:.
- P235-Keep cool.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : N, N, N', N'-tetramethylethylenediamine
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'