Learn More
SI Analytics™ Temperature-Controlled Titration Vessel
SI Analytics™ Temperature-Controlled Titration Vessel is constructed of double jacket glass and includes a removable head.
Brand:  SI Analytics™ 285216060
Product Code. 10197949
  • £1295.00 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Temperature-controlled
  • Double jacket glass vessel
  • With removeable head
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Double Jacket Glass
Removable head
6 x NS 14.5 drillings
Titration Vessel
PP/PTFE
30 to 150 mL