Learn More
Grant Instruments™ Temperature Gradient Plate
GRD1 Highly efficient bi-directional temperature gradient system for investigating responses to temperature shifts of seeds, small plants, insects, micro-organisms or any small component or material.
Brand:  Grant Instruments™ GRD1
Product Code. 11460988
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Temperature range: +5 to 30ºC (cold edges)
  • Temperature range: ambient +5 to 45ºC (hot edges)
  • Perspex grid divides working area into 196 mini-incubators
  • Multi-channel Squirrel data logger (included as standard) for recording time and temperature
  • Over temperature protection is provided on each edge by fixed temperature cut-outs
Specifications
Specifications
5°C to 30°C (cold edges), Ambient +5° to 45°C (hot edges)
±-0.5°C
229 kg
1020D x 1020W x 1040 mmH
760 x 760 mm