Learn More
TFA Dostmann Temperature/Humidity Transmitter
Temperature transmitter for TFA wireless thermo-hygrometers and weather stations.
Brand:  TFA Dostmann 30.3180.IT
Product Code. 17701218
  • £20.70 / Each
Explore more special offers
Description
Description
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3039, 30.3060
  • For wireless transmission of the outdoor climate
  • Display with temperature and humidity
  • Range: 100 m in open field
  • Update: every 4 seconds
  • Also for climate monitoring of remote rooms
Specifications
Specifications
Temperature/Humidity Transmitter