Learn More
Eppendorf™ Temperature Sensor Device for Mastercycler
Rapid, easy to use, and suitable for use with Mastercycler PCR cyclers.
£923.68 - £1054.20
Specifications
Content And Storage | Requires Temperature Sensor |
---|---|
Description | Single Channel |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15733145
|
Eppendorf™
0056001000 |
Mastercycler, 96-well |
£1054.20
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15753145
|
Eppendorf™
0056002006 |
Mastercycler, 384-well |
£923.68
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Measuring device without temperature sensor
- Sensor for 96 well and 384 well blocks, sold separately
- Simple, rapid verification and adjustment
- Automatic adjustment with Mastercycler software, version 2.10 or higher
- Can be third-party adjusted
Specifications
Requires Temperature Sensor | |
Single Channel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Eppendorf™ Temperature Sensor Device for Mastercycler