Learn More
Labconco™ Temperature Sensor for FreeZone™
This Temperature Sensor for FreeZone™ is an RTD sensor for monitoring product sample temperature.
Brand:  Labconco™ 7515300
Additional Details : Weight : 0.45000kg
Product Code. 15431455
  • £142.00 / Each
Estimated Shipment: 02-10-2024
to see stock.
Add to basket
Description
Description
  • Replacement temperature sensor for all Labconco™ FreeZone™ Freeze Dryers
Specifications
Specifications
Sensor
RTD
Labconco™ FreeZone™ Freeze Dryers