Learn More
TFA Dostmann Temperature Transmitter
Temperature transmitter for TFA wireless thermo-hygrometers and weather stations.
Brand:  TFA Dostmann 30.3181.IT
Product Code. 12636967
  • £19.20 / Each
Explore more special offers
Description
Description
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3039, 30.3053, 30.3060
  • For wireless transmission of the outdoor temperature
  • With temperature display
  • With additional cable sensor
  • Update: every 4 seconds
  • Range: 100 m in open field
  • Also for temperature monitoring in remote rooms
Specifications
Specifications
Temperature/Humidity Transmitter