Learn More
TFA Dostmann Temperature Transmitter
Temperature transmitter for TFA wireless thermo-hygrometers and weather stations.
Brand:  TFA Dostmann 30.3147.IT
Product Code. 15321852
  • £13.50 / Each
Explore more special offers
Description
Description
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3009, 30.3016, 30.3018, 30.3022, 30.3030, 30.3032, 30.3037, 35.1026, 35.1044, 35.1048, 35.1064, 35.1068, 35.1076, 35.1084, 35.1089, 35.1115, 35.1117, 35.1125, 98.1006
  • For wireless transmission of the outdoor temperature
  • Also for temperature monitoring in remote rooms
  • Range: 100 m in open field
Specifications
Specifications
Temperature/Humidity Transmitter