Learn More
TFA Dostmann Temperature Transmitter
Transmitter for TFA wireless thermo-hygrometers and weather stations.
Brand:  TFA Dostmann 30.3228.02
Product Code. 16661246
  • £10.85 / Each
Explore more special offers
Description
Description
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3054, 30.3056, 30.3062
  • For wireless transmission of the outdoor temperature
  • With temperature display
  • Range: 100 m in open field
  • Adjustable for channel 1-8
  • Also for temperature monitoring in remote rooms
Specifications
Specifications
Temperature/Humidity Transmitter