missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Tempered cast iron Tripods
£19.65 - £24.05
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10085150
|
Bochem™
5230 |
100 mm |
£19.65
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
10794723
|
Bochem™
5232 |
140 mm |
£24.05
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
Specifications
Tempered Cast Iron | |
Tripods | |
1 Pk. |