Learn More
TEMPO, free radical, 98+%, Thermo Scientific Chemicals
Description
TEMPO is used as a catalyst in organic synthesis and for the oxidation of primary alcohols to aldehydes. It finds use in the chemical industry for conversion of bisnoralcohol (a steroid) to bisnoraldehyde. It acts as a free radical scavenger, as a mediator in controlled radical polymerization and as a structural probe in electron spin resonance spectroscopy. Further, it is involved in the preparation of (S)-(+)-2-methylbutanal from (S)-(-)-2-methyl-1-butanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TEMPO is used as a catalyst in organic synthesis and for the oxidation of primary alcohols to aldehydes. It finds use in the chemical industry for conversion of bisnoralcohol (a steroid) to bisnoraldehyde. It acts as a free radical scavenger, as a mediator in controlled radical polymerization and as a structural probe in electron spin resonance spectroscopy. Further, it is involved in the preparation of (S)-(+)-2-methylbutanal from (S)-(-)-2-methyl-1-butanol.
Solubility
Soluble in all organic solvents. Insoluble in water.
Notes
Store in a cool place. Incompatible withstrong oxidizing agents and strong acids.
Specifications
Specifications
Melting Point | 31°C to 40°C |
Boiling Point | 193°C (decomposition) |
Flash Point | 67°C (152°F) |
Quantity | 1 g |
UN Number | UN3263 |
Beilstein | 1422418 |
Merck Index | 14,9140 |
Solubility Information | Soluble in all organic solvents. Insoluble in water. |
Formula Weight | 156.25 |
Percent Purity | ≥98% |
Show More |
Safety and Handling
- TEMPO, free radical
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 2,2,6,6-Tetramethylpiperidinooxy
RUO – Research Use Only