Learn More
Tocris Bioscience™ Temsirolimus
mTOR inhibitor; antitumor
Brand:  Tocris Bioscience™ 5264/10
Product Code. 18775298
  • £168.00 / 10mg
Explore more special offers
Description
Description
Temsirolimus is a mTOR inhibitor; inhibits tumor growth in breast cancer cell lines (IC50 values are 1.6 and 4.3 nM for SKBr3 and BT474, respectively). Inhibits HIF-1α-mediated VEGF production in breast cancer cell lines (BT474 and MDA-MB-231). Directly inhibits serum and VEGF mediated endothelial cell proliferation and morphogenesis in vitro and vessel formation in vivo. Causes G1/S cell cycle arrest in multiple cancer cell lines. Antiangiogenic. Activates autophagy.
Specifications
Specifications
>98%
10 mg
1030.29
CCI-779
Temsirolimus
Temsirolimus