Learn More
Cole-Parmer™ Tensiometer, Capillary Surface Tension Apparatus
Quickly and easily determine surface tension based on height of liquid in capillary tube
Brand: Cole-Parmer™ 59780-90
Code : NEW
Additional Details : Weight : 0.27000kg
Description
- Clear, easy-to-read borosilicate glass capillary tube
- Included rubber stopper centers glass capillary tube in the outer cylinder
- Apparatus is comprised of an engraved 250-mm borosilicate glass capillary tube (graduated from 0 to 10 cm in 1-mm divisions) and an outer tube.
- The capillary tube is positioned in the center of the outer tube by means of a rubber stopper.
- The outer tube accommodates a pipette bulb to expel air.
Specifications
Tensiometer, Capillary Surface Tension Apparatus | |
250 mm borosilicate glass capillary tube, rubber stopper, and outer cylinder |