missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TentaGel™ S-NH2, Thermo Scientific Chemicals
£130.00 - £581.00
Specifications
Chemical Name or Material | TentaGel™ S-NH2 |
---|---|
Color | White to Yellow |
Physical Form | Powder |
Loading | 0.2 to 0.3mmol/g |
Particle Size | 90 Micron |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10246161
|
Thermo Scientific Acros
354610050 |
5 g | Glass bottle |
£130.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317911
|
Thermo Scientific Acros
354610250 |
25 g | Glass Bottle |
£581.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesisSpecifications
TentaGel™ S-NH2 | |
Powder | |
90 Micron | |
O-(2-Aminoethyl)polyethylene glycol polymer bound |
White to Yellow | |
0.2 to 0.3mmol/g | |
MFCD00217899 |
RUO – Research Use Only