Learn More
Hamilton™ Terasaki Dispenser
This multi-syringe dispenser accurately dispenses six samples simultaneously up to 50 times with the press of a lever.
Brand: Hamilton™ 201920
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
- Uses removable needles
- Available for a range of volumes
Specifications
1/Pk. | |
Terasaki Dispenser | |
100 μL |