missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbinafine hydrochloride
Selective TASK3 activator
£43.00 - £135.00
Specifications
CAS | 78628-80-5 |
---|---|
Synonym | N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride |
Molecular Formula | C21H25N.HCl |
Formula Weight | 327.89 |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18716188
|
Tocris Bioscience
6484/50 |
50 mg |
£43.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760878
|
Tocris Bioscience
6484/250 |
250 mg |
£135.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective TASK3 activator (pEC50 = 6.2). Exhibits >10-fold selectivity for TASK3 over TREK2, TRESK, THIK1 and TASK2. Also inhibits TWIK1 (pIC50 = 5.69). Also antifungal; inhibits fungal sterol biosynthesis by inhibition of squalene epoxidase.Specifications
78628-80-5 | |
C21H25N.HCl | |
98% |
N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride | |
327.89 |
Safety and Handling
Recommended Storage : Store at 4°C