missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) 2,4-pentanedionate, 99.9% (REO)

Product Code. 11369217
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11369217

Brand: Thermo Scientific Alfa Aesar 040032.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14284-95-8 | C15H21O6Tb | 456.25 g/mol

Terbium(III) 2,4-pentanedionate is used as an active pharmaceutical ingredient. It is also used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-95-8
Molecular Formula C15H21O6Tb
Molecular Weight (g/mol) 456.25
MDL Number MFCD00013503
InChI Key MAPWZFZLNRMAMO-UHFFFAOYSA-N
Synonym terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11
PubChem CID 129878174
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;terbium
SMILES [Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 456.25
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Terbium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.