missing translation for 'onlineSavingsMsg'
Learn More
Terbium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11358208

Terbium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11358208
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11358208

Brand: Thermo Scientific Alfa Aesar 039425.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14456-47-4
Molecular Formula Br3Tb
Molecular Weight (g/mol) 398.637
MDL Number MFCD00049571
InChI Key AZNZWHYYEIQIOC-UHFFFAOYSA-K
Synonym terbium iii bromide, terbium tribromide, terbium 3+ tribromide, terbium iii ; bromide, acmc-1btzk, terbium 3+ ion tribromide, terbium bromideShow MoreShow Less
PubChem CID 84452
IUPAC Name terbium(3+);tribromide
SMILES [Br-].[Br-].[Br-].[Tb+3]

Specifications

Boiling Point 1490°C
Melting Point 827°C
Physical Form Powder
Quantity 5 g
Formula Weight 398.65
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) bromide, Ultra dry

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM30K024 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number039425.03,039425.06
Lot NumberX11J042 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number039425.03,039425.06
Lot NumberF15U041 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039425.03,039425.06
Lot NumberL11L31 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number039425.03,039425.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.