missing translation for 'onlineSavingsMsg'
Learn More
Terbium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11386248

Terbium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11386248
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11386248

Brand: Thermo Scientific Alfa Aesar 011209.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Terbium(III) chloride hexahydrate is a water soluble source used for the preparation of metallic terbium, which is used in electronic devices and an additive for special alloy namely Terfenol-D. It is used in the preparation of complexes, which are used in the spectrofluorimetric determination of various chemicals in pharmaceutical formulations. Further, it finds use in the preparation of terbium nanotubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13798-24-8
Molecular Formula Cl3H12O6Tb
Molecular Weight (g/mol) 373.37
MDL Number MFCD00149870
InChI Key ULJUVCOAZNLCJZ-UHFFFAOYSA-K
IUPAC Name terbium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.9% (REO)
Solubility Information Very soluble in water.
Formula Weight 373.38 (265.29 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Terbium(III) chloride hexahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX09K005 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog Number011209.18,011209.09
Lot NumberW07K004 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog Number011209.18,011209.09
Lot NumberS10I008 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number011209.18,011209.09
Lot NumberR26H016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011209.18,011209.09
Lot NumberA29Z037 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011209.18,011209.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.