missing translation for 'onlineSavingsMsg'
Learn More
Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11396908

Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11396908
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11396908

Brand: Thermo Scientific Alfa Aesar 013657.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Terbium(III) fluoride is used as a source of terbium. It is also useful in the preparation of fluoride glasses. Further, it is used in the preparation of electroluminescent thin films and luminescent zinc sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13708-63-9
Molecular Formula F3Tb
Molecular Weight (g/mol) 215.92
MDL Number MFCD00049572
InChI Key LKNRQYTYDPPUOX-UHFFFAOYSA-K
IUPAC Name terbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tb+3]

Specifications

Melting Point 1172°C
Physical Form Crystalline
Quantity 5 g
UN Number UN3288
Solubility Information Slightly soluble in mineral acids. Insoluble in water.
Formula Weight 215.92
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) fluoride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX18I039 Certificate TypeCertificate of Analysis Date26/10/2022 Catalog Number013657.06,013657.14
Lot NumberT16G033 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number013657.06,013657.14
Lot NumberP3555A Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number013657.06,013657.14
Lot NumberX02A024 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number013657.06,013657.14
Lot NumberG19X004 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number013657.06,013657.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.