Learn More
Terbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
£24.11 - £365.00
Chemical Identifiers
CAS | 13708-63-9 |
---|---|
Molecular Formula | F3Tb |
Molecular Weight (g/mol) | 215.92 |
MDL Number | MFCD00049572 |
InChI Key | LKNRQYTYDPPUOX-UHFFFAOYSA-K |
IUPAC Name | terbium(3+) trifluoride |
SMILES | [F-].[F-].[F-].[Tb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326258
|
Thermo Scientific Alfa Aesar
011213.03 |
1 g | N/A | ||||||
11346258
|
Thermo Scientific Alfa Aesar
011213.06 |
5 g |
£112.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336258
|
Thermo Scientific Alfa Aesar
|
25 g |
£365.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Terbium(III) fluoride is used as a source of terbium. It is also useful in the preparation of fluoride glasses. Further, it is used in the preparation of electroluminescent thin films and luminescent zinc sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13708-63-9 | |
215.92 | |
LKNRQYTYDPPUOX-UHFFFAOYSA-K | |
[F-].[F-].[F-].[Tb+3] |
F3Tb | |
MFCD00049572 | |
terbium(3+) trifluoride |
Specifications
1172°C | |
Crystalline | |
MFCD00049572 | |
Slightly soluble in mineral acids. Insoluble in water. | |
[F-].[F-].[F-].[Tb+3] | |
215.92 | |
99.99% | |
Odorless | |
(REO) |
13708-63-9 | |
F3Tb | |
UN3288 | |
LKNRQYTYDPPUOX-UHFFFAOYSA-K | |
terbium(3+) trifluoride | |
215.92 | |
Reacton™ | |
Hygroscopic | |
Terbium(III) fluoride, Anhydrous |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 237-247-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only