Learn More
Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
POA
Specifications
Melting Point | 2340°C |
---|---|
CAS | 12037-01-3 |
Physical Form | Solid |
Molecular Formula | Tb{4}O{7} |
Percent Purity | 99.9% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11336248
|
Thermo Scientific Alfa Aesar
011207.30 |
250 g |
POA
250g |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11346248
|
Thermo Scientific Alfa Aesar
011207.A1 |
1 kg |
POA
1kg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
Terbium(III,IV) oxide acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optical and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2340°C | |
Solid | |
99.9% |
12037-01-3 | |
Tb{4}O{7} |