missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III,IV) oxide, REacton™, 99.99% (REO)

Product Code. 11366248
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11366248

Brand: Thermo Scientific Alfa Aesar 011208.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12037-01-3 | O7Tb4 | 747.69 g/mol

Terbium(III,IV) oxide is used in the preparation of other terbium compounds. Activating agents for fluorescent powders, composites for catalytic automobile exhaust converters, Glass materials with a Faraday Rotation effect for optical and laser-based devices, Magneto-optical glasses, Magneto-optical recording materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-01-3
Molecular Formula O7Tb4
Molecular Weight (g/mol) 747.69
MDL Number MFCD00083151
InChI Key UWZVBPNIJCKCMC-UHFFFAOYSA-N
IUPAC Name tetraoxotetraterboxane
SMILES O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O

Specifications

Melting Point 2340°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information soluble in dilute acids. Insoluble in water.
Formula Weight 747.69
Chemical Name or Material Terbium(III, IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.