missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) nitrate hexahydrate, 99.99+%, (trace metal basis)

Product Code. p-221660 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 15249164

Brand: Thermo Scientific Acros 295790020

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13451-19-9 | H12N3O15Tb | 453.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13451-19-9
Molecular Formula H12N3O15Tb
Molecular Weight (g/mol) 453.03
MDL Number MFCD00799162,MFCD00149868
InChI Key UTCARTSNNKGRTD-UHFFFAOYSA-N
Synonym terbium trinitrate hexahydrate, terbium iii nitrate hexahydrate, terbium nitrate, hexahydrate, terbium iii nitrate, hexahydrate 1:3:6, terbium nitrate hexahydrate, nitric acid, terbium 3+ salt, hexahydrate, 3no3.tb.6h2o, terbium iii nitratehexahydrate, terbium 3+ trinitrate hexahydrate, terbium iii nitrate hydrate, reacton
PubChem CID 202879
IUPAC Name terbium(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Physical Form Granular Powder
Quantity 2 g
Assay Percent Range 99.99% min. (TREO base)
Linear Formula Tb(NO3)3·6H2O
Beilstein 11,9087
Merck Index 14,9157
Solubility Information Solubility in water: soluble
Formula Weight 453.02
Percent Purity ≥99.99%
Grade Trace Metal Basis
Chemical Name or Material Terbium(III) nitrate hexahydrate, 99.9%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Terbium(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.