missing translation for 'onlineSavingsMsg'
Learn More
Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11342548

Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11342548
10 g
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11342548

Brand: Thermo Scientific Alfa Aesar 014596.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13842-67-6
MDL Number MFCD00149872

Specifications

Melting Point 360°C (in -8H2O)
Physical Form Crystalline
Quantity 10 g
Formula Weight 750.19 (606.05 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Assay (REO)
Chemical Name or Material Terbium(III) sulfate octahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS20K035 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number014596.09,014596.18,014596.04
Lot NumberN19K017 Certificate TypeCertificate of Analysis Date15/05/2024 Catalog Number014596.09,014596.18,014596.04
Lot NumberZ12J006 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number014596.09,014596.18,014596.04
Lot NumberZ15I013 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number014596.09,014596.18,014596.04
Lot NumberR27I022 Certificate TypeCertificate of Analysis Date07/06/2022 Catalog Number014596.09,014596.18,014596.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.