missing translation for 'onlineSavingsMsg'
Learn More
Terbium pieces, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11336527

Terbium pieces, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11336527
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11336527

Brand: Thermo Scientific Alfa Aesar 000324.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Physical Form Pieces
Quantity 5 g
Assay Percent Range 99.9% (REO)
Odor Odorless
Appearance Silver Gray pieces
Chemical Name or Material Terbium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX07K015 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number000324.03,000324.06
Lot NumberY05H005 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000324.03,000324.06
Lot NumberA10L30 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000324.03,000324.06
Lot NumberG07T007 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000324.03,000324.06
Lot Number61700654 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000324.03,000324.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium pieces, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.