missing translation for 'onlineSavingsMsg'
Learn More

Terbium, plasma standard solution, Specpure™ Tb 1000μg/mL, Thermo Scientific Chemicals

£64.92 - £206.03

Chemical Identifiers

CAS 12037-01-3, 7697-37-2, 7732-18-5
Molecular Formula O7Tb4
Molecular Weight (g/mol) 747.69
MDL Number MFCD00083151
InChI Key UWZVBPNIJCKCMC-UHFFFAOYSA-N
IUPAC Name tetraoxotetraterboxane
SMILES O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361429
View Documents
Thermo Scientific Alfa Aesar
013860.AE
100 mL
£64.92
100mL
Please to purchase this item. Need a web account? Register with us today!
11490360
View Documents
Thermo Scientific Alfa Aesar
013860.AP
500 mL
£206.03
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12037-01-3, 7697-37-2, 7732-18-5
747.69
UWZVBPNIJCKCMC-UHFFFAOYSA-N
O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
O7Tb4
MFCD00083151
tetraoxotetraterboxane

Specifications

12037-01-3, 7697-37-2, 7732-18-5
MFCD00083151
O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
747.69
23 hPa (17mm Hg) at 20°C
Tellurium
O7Tb4
UWZVBPNIJCKCMC-UHFFFAOYSA-N
tetraoxotetraterboxane
Nitric acid: 5%; Terbium(III, IV) oxide: 0.12%; Water: 94.88%
Liquid
Videos
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III

missing translation for 'einecsNumber' : 231-714-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.