missing translation for 'onlineSavingsMsg'
Learn More

Terephthalamidoxime, 97%, Thermo Scientific™

£36.40 - £152.00

Chemical Identifiers

CAS 6051-62-3
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.194
MDL Number MFCD00174328
InChI Key JUXCYRYHZGAKKG-UHFFFAOYSA-N
Synonym terephthalamidoxime, n1,n4-dihydroxyterephthalimidamide, 1,4-diamidoximobenzene, n'1,n'4-dihydroxybenzene-1,4-dicarboximidamide, terephthaldiamidoxime, 1,4-benzenediamide dioxime, n,4-benzenedicarboximidamide, benzene-1,4-dicarboxamidoxime, grwntbqzuuoolq-uhfffaoysa-n, n1,n4-dihydroxybenzene-1,4-dicarboximidamide
PubChem CID 606612
IUPAC Name N-[amino-[4-[amino(nitroso)methylidene]cyclohexa-2,5-dien-1-ylidene]methyl]hydroxylamine
SMILES C1=CC(=C(N)N=O)C=CC1=C(N)NO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15498807
View Documents
Thermo Scientific Alfa Aesar
H52221.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15408817
View Documents
Thermo Scientific Alfa Aesar
H52221.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6051-62-3
194.194
JUXCYRYHZGAKKG-UHFFFAOYSA-N
606612
C1=CC(=C(N)N=O)C=CC1=C(N)NO
C8H10N4O2
MFCD00174328
terephthalamidoxime, n1,n4-dihydroxyterephthalimidamide, 1,4-diamidoximobenzene, n'1,n'4-dihydroxybenzene-1,4-dicarboximidamide, terephthaldiamidoxime, 1,4-benzenediamide dioxime, n,4-benzenedicarboximidamide, benzene-1,4-dicarboxamidoxime, grwntbqzuuoolq-uhfffaoysa-n, n1,n4-dihydroxybenzene-1,4-dicarboximidamide
N-[amino-[4-[amino(nitroso)methylidene]cyclohexa-2,5-dien-1-ylidene]methyl]hydroxylamine

Specifications

6051-62-3
MFCD00174328
JUXCYRYHZGAKKG-UHFFFAOYSA-N
N-[amino-[4-[amino(nitroso)methylidene]cyclohexa-2,5-dien-1-ylidene]methyl]hydroxylamine
606612
97%
C8H10N4O2
terephthalamidoxime, n1,n4-dihydroxyterephthalimidamide, 1,4-diamidoximobenzene, n'1,n'4-dihydroxybenzene-1,4-dicarboximidamide, terephthaldiamidoxime, 1,4-benzenediamide dioxime, n,4-benzenedicarboximidamide, benzene-1,4-dicarboxamidoxime, grwntbqzuuoolq-uhfffaoysa-n, n1,n4-dihydroxybenzene-1,4-dicarboximidamide
C1=CC(=C(N)N=O)C=CC1=C(N)NO
194.194
194.19
Terephthalamidoxime
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications