Learn More
Thermo Scientific™ Tergitol™ 7-Lactose-TTC Agar
Enumerate and differentiate coliforms from water and food samples with this ready-to-use medium.
£14.45 - £18.10
Specifications
Color | Reseda green, transparent |
---|---|
Content And Storage | 2° to 12°C, in the dark |
Description | Tergitol 7-Lactose-TTC Agar |
Format | 90mm Plate |
Manufacturing Origin | Germany |
Product Code | Brand | Packaging Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Price | Quantity & Availability | |||||
10670795
|
Thermo Scientific™
PO5164A |
Flow wrapped - Single Layer |
£14.45
Pack of 10 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
11908429
|
Thermo Scientific™
PO5411J |
Boxes with 2 x 10 plates wrapped in film |
£18.10
Pack of 10 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Description
Enumerate and differentiate coliforms from water and food samples with ready-to-use Thermo Scientific™ Oxoid™ Tergitol™ 7-Lactose-TTC Agar. Tergitol 7 inhibits Gram-positive organisms allowing superior recovery of coliforms. The addition of tri-phenyltetrazolium chloride (TTC) allows recognition and identification of Escherichia coli and Enterobacter aerogenes. Fermentation of lactose is seen by a change in colour of the pH indicator bromothymol blue. TTC is rapidly reduced to red formazan by most coliform organisms except E. coli and Enterobacter aerogenes allowing easy differentiation.
Use Tergitol 7-Lactose-TTC Agar for the detection and enumeration of coliforms in food and water samples.
- Easy-to-read: easily differentiate Escherichia coli (yellow colonies) and Enterobacter species (greenish/yellow colonies) from coliforms (red colonies).
- Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Tergitol™ is a trademark of the Dow Chemical Company (Dow)
Specifications
Reseda green, transparent | |
Tergitol 7-Lactose-TTC Agar | |
Germany |
2° to 12°C, in the dark | |
90mm Plate | |
Agar |
Safety and Handling
missing translation for 'shelfLife' : 12 Weeks