Learn More
Fisher BioReagents™ Terrific Broth
Fisher BioReagents™ Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E.coli.
Brand: Fisher BioReagents™ 12891660
Code : 32
Additional Details : Weight : 1.80000kg
Description
- Modified large granules
Specifications
Terrific Broth | |
Microbiology Media | |
2000 g |