missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Terrific Broth, Modified (Granulated)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
£68.26 - £268.72
Specifications
Form | Granulated Powder |
---|---|
Grade | Molecular genetics |
Media Type | Terrific Broth |
Product Type | Microbiology Media |
Formulation | Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11306002
|
Fisher BioReagents™
BP9728-500 |
Poly Bottle | 500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11395992
|
Fisher BioReagents™
BP9728-2 |
Poly Bottle | 2 kg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Directions for Media Preparation: Add 47.6g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Then, add 4 mL of glycerol to the mixture. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Greater peptone and yeast concentration allow for higher plasmid yield
- Format: Granulated powder
Specifications
Granulated Powder | |
Terrific Broth | |
Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L |
Molecular genetics | |
Microbiology Media | |
For maintenance and propagation of Escherichia coli in molecular biology procedures |