missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Terrific Broth (Powder/Molecular Genetics)

Enriched medium used with glycerol in cultivating recombinant strains of Escherichia coli.

£32.40 - £1547.00

Specifications

Content And Storage 4°C
Form Powder
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
View More Specs

Products 4
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
10041363
View Documents
Fisher BioReagents™
BP2468-500
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10173243
View Documents
Fisher BioReagents™
BP2468-100
100 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10513325
View Documents
Fisher BioReagents™
BP2468-2
2000 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10583145
View Documents
Fisher BioReagents™
BP2468-10
10,000 g Poly Pail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Formulation Usage: 50.8g/L

Specifications

Specifications

4°C
Molecular genetics
Microbiology Media
Powder
Terrific Broth
Cultivation of recombinant strains of Escherichia coli
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Broth Miller

SDS
Documents

Documents

Product Certifications