missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Terry Heavy Gloves
Click to view available options
For use with: Automotive, Glass, Iron, Steel industries
Description
- For mechanical and thermal handlings in glassworks, metallurgy and foundry.
- 100% cotton terry glove.
- 5-finger glove for a better grip on hot objects and added dexterity.
- Straight thumb.
- The cuff helps to protect a large part of the forearm.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-04 Thermical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/06/08/0184, EC Attestation Number 0075/797/162/06/08/0184 |
Classification | Dexterity: 5 |
Color | Beige |
Cut Resistance Rating | 3 |
Material | Terrycloth, Cotton |
Product Type | Gloves |
Temperature (Metric) Max | 250°C |
For Use With (Application) | Automotive, Glass, Iron, Steel industries |
Puncture Rating | 1 |
Quantity | 6Pair/PK |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'