Learn More
Thermo Scientific™ Terry Tool Tubing Cutters
Shop All Thermo Scientific ProductsDescription
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
Specifications
Specifications
| Type | Tubing Cutter |
| Description | Cuts 0.125 stainless steel tubing |
| For Use With (Equipment) | 1/8 in. Stainless Steel Tubing |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.