missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl 2,2,2-trichloroacetimidate, 95%, Thermo Scientific Chemicals Product Code.: 10767403

tert-Butyl 2,2,2-trichloroacetimidate, 95%, Thermo Scientific Chemicals

Product Code. 10767403
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10767403

Brand: Thermo Scientific Acros 369330050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98946-18-0
Molecular Formula C6H10Cl3NO
Molecular Weight (g/mol) 218.50
MDL Number MFCD00077410
InChI Key CQXDYHPBXDZWBA-UHFFFAOYSA-N
Synonym tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimineShow MoreShow Less
PubChem CID 2734700
IUPAC Name tert-butyl 2,2,2-trichloroethanimidate
SMILES CC(C)(C)OC(=N)C(Cl)(Cl)Cl

Specifications

Melting Point 20.0°C
Color Colorless to Yellow
Density 1.2220g/mL
Boiling Point 64.0°C to 66.0°C (15.0 mmHg)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CCl3C(=NH)OC(CH3)3
Refractive Index 1.4550 to 1.4570
Quantity 5 g
Specific Gravity 1.222
Formula Weight 218.51
Percent Purity 95%
Physical Form Liquid or Low Melting Crystalline Mass
Chemical Name or Material tert-Butyl 2, 2, 2-trichloroacetimidate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0414521 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number369330050,369330250,369331000
Lot NumberA0459284 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number369330050,369330250,369331000
Lot NumberA0393030 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number369330050,369330250,369331000
Lot NumberA0409866 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number369330050,369330250,369331000
Lot NumberA0386458 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number369330050,369330250,369331000
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl 2,2,2-trichloroacetimidate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 2,2,2-trichloroacetimidate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.