missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2,2,2-trichloroacetimidate, 97%, Thermo Scientific Chemicals

Product Code. 11479527
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

tert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.

Solubility
Soluble in organic solvents, cyclo hexane.

Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98946-18-0
Molecular Formula C6H10Cl3NO
Molecular Weight (g/mol) 218.50
MDL Number MFCD00077410
InChI Key CQXDYHPBXDZWBA-UHFFFAOYSA-N
PubChem CID 2734700
IUPAC Name tert-butyl 2,2,2-trichloroethanimidate
SMILES CC(C)(C)OC(=N)C(Cl)(Cl)Cl

Specifications

Melting Point 20°C to 22°C
Density 1.222
Boiling Point 64°C to 66°C (15 mmHg)
Flash Point 55°C (131°F)
Assay Percent Range 0.97
Refractive Index 1.457
Quantity 5 g
UN Number UN1993
Beilstein 1770049
Sensitivity Moisture sensitive
Synonym tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine
Solubility Information Soluble in organic solvents,cyclo hexane.
Formula Weight 218.51
Percent Purity 97%
Chemical Name or Material tert-Butyl 2,2,2-trichloroacetimidate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'