missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-bromo-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals

£262.64 - £948.00

Chemical Identifiers

CAS 365996-06-1
Molecular Formula C11H15BrN2O2S
Molecular Weight (g/mol) 319.22
InChI Key RXXKNZKFRAFFOH-UHFFFAOYSA-N
Synonym tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyridine-5 4h-carboxylate, 5-boc-2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine, tert-butyl 2-bromo-6,7-dihydro 1,3 thiazolo 5,4-c pyridine-5 4h-carboxylate, tert-butyl 2-bromo-4h,5h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, 2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylic acid tert-butyl ester, 2-bromo-4,5,6,7-tetrahydro 1,3 thiazolo 5,4-c pyridine, n-boc protected, tert-butyl 2-bromo-4h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, pubchem15507, tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyrid, 5-n-boc-2-bromo-4,5,6,7-tetrahydrothiazolo 5,4-c pyridine
PubChem CID 11162951
IUPAC Name tert-butyl 2-bromo-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2=C(C1)SC(=N2)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15244308
View Documents
Thermo Scientific Acros
453590010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15254308
View Documents
Thermo Scientific Acros
453590050
5 g
£948.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

365996-06-1
319.22
tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyridine-5 4h-carboxylate, 5-boc-2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine, tert-butyl 2-bromo-6,7-dihydro 1,3 thiazolo 5,4-c pyridine-5 4h-carboxylate, tert-butyl 2-bromo-4h,5h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, 2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylic acid tert-butyl ester, 2-bromo-4,5,6,7-tetrahydro 1,3 thiazolo 5,4-c pyridine, n-boc protected, tert-butyl 2-bromo-4h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, pubchem15507, tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyrid, 5-n-boc-2-bromo-4,5,6,7-tetrahydrothiazolo 5,4-c pyridine
tert-butyl 2-bromo-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
C11H15BrN2O2S
RXXKNZKFRAFFOH-UHFFFAOYSA-N
11162951
CC(C)(C)OC(=O)N1CCC2=C(C1)SC(=N2)Br

Specifications

365996-06-1
Authentic
Glass Bottle
tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyridine-5 4h-carboxylate, 5-boc-2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine, tert-butyl 2-bromo-6,7-dihydro 1,3 thiazolo 5,4-c pyridine-5 4h-carboxylate, tert-butyl 2-bromo-4h,5h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, 2-bromo-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylic acid tert-butyl ester, 2-bromo-4,5,6,7-tetrahydro 1,3 thiazolo 5,4-c pyridine, n-boc protected, tert-butyl 2-bromo-4h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, pubchem15507, tert-butyl 2-bromo-6,7-dihydrothiazolo 5,4-c pyrid, 5-n-boc-2-bromo-4,5,6,7-tetrahydrothiazolo 5,4-c pyridine
CC(C)(C)OC(=O)N1CCC2=C(C1)SC(=N2)Br
319.22
319.22
tert-Butyl 2-bromo-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate
94.0°C to 97.0°C
96% min. (HPLC)
C11H15BrN2O2S
RXXKNZKFRAFFOH-UHFFFAOYSA-N
tert-butyl 2-bromo-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
11162951
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

Product Identifier
  • tert-Butyl 2-bromo-6,7-dihydro¢1,3!thiazolo¢5,4-c!pyridine-5(4H)-carboxylate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only