missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 3-bromo-2-furoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 247.09 |
Percent Purity | 97% |
Chemical Name or Material | tert-Butyl 3-bromo-2-furoate |
Safety and Handling
Product Identifier
- tert-Butyl 3-bromo-2-furoate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'