missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific Chemicals

£27.50 - £663.00

Chemical Identifiers

CAS 145549-76-4
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.208
MDL Number MFCD12827541
InChI Key JINYZTGTQXDUQR-UHFFFAOYSA-N
Synonym tert-butyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid tert-butyl ester, cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, tert-butyl 3-oxo-cyclobutanecarboxylate, 1-boc-cyclobutan-3-one, t-butyl 3-oxocyclobutanecarboxylate, 3-tert-butyloxycarbonylcyclobutanone, tert-butyl3-oxocyclobutanecarboxylate, tert-butyl 3-oxocyclobutane carboxylate, t-butyl-3-oxocyclobutanecarboxylate
PubChem CID 10261520
IUPAC Name tert-butyl 3-oxocyclobutane-1-carboxylate
SMILES CC(C)(C)OC(=O)C1CC(=O)C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418658
View Documents
Thermo Scientific Alfa Aesar
H62064.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15498648
View Documents
Thermo Scientific Alfa Aesar
H62064.03
1 g
£157.79
1g
Please to purchase this item. Need a web account? Register with us today!
15408658
View Documents
Thermo Scientific Alfa Aesar
H62064.06
5 g
£663.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

145549-76-4
170.208
JINYZTGTQXDUQR-UHFFFAOYSA-N
10261520
CC(C)(C)OC(=O)C1CC(=O)C1
C9H14O3
MFCD12827541
tert-butyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid tert-butyl ester, cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, tert-butyl 3-oxo-cyclobutanecarboxylate, 1-boc-cyclobutan-3-one, t-butyl 3-oxocyclobutanecarboxylate, 3-tert-butyloxycarbonylcyclobutanone, tert-butyl3-oxocyclobutanecarboxylate, tert-butyl 3-oxocyclobutane carboxylate, t-butyl-3-oxocyclobutanecarboxylate
tert-butyl 3-oxocyclobutane-1-carboxylate

Specifications

145549-76-4
MFCD12827541
JINYZTGTQXDUQR-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutane-1-carboxylate
10261520
97%
C9H14O3
tert-butyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid tert-butyl ester, cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, tert-butyl 3-oxo-cyclobutanecarboxylate, 1-boc-cyclobutan-3-one, t-butyl 3-oxocyclobutanecarboxylate, 3-tert-butyloxycarbonylcyclobutanone, tert-butyl3-oxocyclobutanecarboxylate, tert-butyl 3-oxocyclobutane carboxylate, t-butyl-3-oxocyclobutanecarboxylate
CC(C)(C)OC(=O)C1CC(=O)C1
170.208
170.2
tert-Butyl 3-oxocyclobutanecarboxylate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only