missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-(thiocarbamoyl)piperidine-1-carboxylate, 95%, Thermo Scientific™

Product Code. 10672275
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 274682-80-3
Molecular Formula C11H20N2O2S
Molecular Weight (g/mol) 244.353
MDL Number MFCD10700060
InChI Key ZCELPXRDWJCIJE-UHFFFAOYSA-N
Synonym tert-butyl 3-thiocarbamoyl piperidine-1-carboxylate, 1-tert-butoxycarbonyl piperidine-3-thiocarboxamide, 3-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl-3-carbamothioylpiperidine, 3-carbamothioylpiperidine, n1-boc protected, 3-carbamothioylpiperidine,n1-boc protected, tert-butyl3-carbamothioylpiperidine-1-carboxylate, tert-butyl 3-aminothioxomethyl piperidinecarboxylate, tert-butyl 3-aminocarbonothioyl piperidine-1-carboxylate, 1-piperidinecarboxylicacid, 3-aminothioxomethyl-, 1,1-dimethylethyl ester
PubChem CID 23546083
IUPAC Name tert-butyl 3-carbamothioylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=S)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 244.36
Percent Purity 95%
Chemical Name or Material Tert-butyl 3-(thiocarbamoyl)piperidine-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.